missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR1 Mouse anti-Human, CoraLite 488, Clone: 1F5D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66923-20UL
Additional Details : Weight : 0.00010kg
Description
ROR1 Monoclonal antibody apecifically detetects ROR1 antigen in Human Samples. It is validated for Flow Cytometry.Specifications
ROR1 | |
Monoclonal | |
1000 ug/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
ROR1 | |
ROR1 fusion protein Ag14586 | |
20 μL | |
4919 | |
Store at -20°C. Avoid exposure to light. | |
IgG1 |
Flow Cytometry | |
1F5D8 | |
CoraLite 488 | |
BC006374 | |
Mouse | |
Protein G purification | |
Primary | |
Human | |
Liquid |