Learn More
Brady™ Rigid Polypropylene First Aid Signs
Brand: Brady™ 810691
Additional Details : Weight : 0.05000kg
Specifications
Emergency Phone Signal | |
NF X 08-003-1 and 08-003-3 NX and the European Directive 92/58 / EEC | |
Polypropylene | |
Rigid Polypropylene | |
120°C | |
10 mm |
Mini Pictograms Hazardous Label | |
Rectangular | |
1 Ea. | |
200 mm | |
0.015 mm |