missing translation for 'onlineSavingsMsg'
Learn More

Rhodanine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7024096
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
141-84-4
C3H3NOS2
133.183
MFCD00005488
KIWUVOGUEXMXSV-UHFFFAOYSA-N
rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazoline
1201546
CHEBI:8830
2-sulfanylidene-1,3-thiazolidin-4-one
C1C(=O)NC(=S)S1
This item is not returnable. View return policy
141-84-4
C3H3NOS2
133.183
MFCD00005488
KIWUVOGUEXMXSV-UHFFFAOYSA-N
rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazoline
1201546
CHEBI:8830
2-sulfanylidene-1,3-thiazolidin-4-one
C1C(=O)NC(=S)S1

Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.

Solubility
Soluble in water (2.25 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 166°C to 171°C
Density 0.868
Odor Odorless
Quantity 25 g
Beilstein 110701
Merck Index 14,8184
Solubility Information Soluble in water (2.25g/L at 20°C).
Formula Weight 133.19
Percent Purity ≥98%
Chemical Name or Material Rhodanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'