Learn More
RETSCH ZM 200 Mill Accessories
Push-fit assembly, with wear-resistant coating
3170.00 EUR
Specifications
For Use With (Equipment) | Ultra Centrifugal mill ZM 200 |
---|---|
Includes | Wear-resistant coating |
Material | Stainless Steel |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
10237940
|
RETSCH
02.608.0044 |
12-tooth Rotor |
3170.00 EUR
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
10370201
|
RETSCH
02.608.0043 |
6-tooth Rotor |
3170.00 EUR
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
Description
For use with ZM 200 Mixer Mill. Choose rotor based on properties of sample, required final fineness and subsequent analysis.
- Stainless-steel, push-fit, with wear-resistant coating
- 6-tooth rotor for coarse, bulky, fibrous goods such as feed pellets, hay and straw
- 12-tooth rotor for medium-coarse goods such as wheat, oats, corn, tablets, powder coatings and plastics
Specifications
Ultra Centrifugal mill ZM 200 | |
Wear-resistant coating | |
Stainless Steel |