Learn More
RETSCH Test Sieves - 14 Mesh or Greater
Stainless steel with wire gauze
Brand: RETSCH 60.150.000180
116.85 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.35000kg
Specifications
Stainless Steel | |
8 in. | |
5.08cm | |
Test Sieve | |
8 in. | |
80 Mesh |
Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc. | |
2 in. | |
180μm | |
ASTM E11 | |
Stainless Steel | |
No. 80 |