Learn More
Trajan™ Replacement Needles for Luer-Lock Syringes: Bevel
Bevel tip style. Trajan Scientific and Medical™ Replacement Needles for Luer-Lock Syringes are supplied in packs of 5.
Brand:  Trajan™ 039862
Additional Details : Weight : 0.80000kg
Product Code. 11939427
  • 63.40 EUR / Pack of 5
Estimated Shipment: 15-07-2024
Description
  • Needle length is the exposed length of needle
  • Bevel tip style
  • Supplied in packs of 5
Specifications
Specifications
16 G
1.1 mm
1.57 mm
Needle