Learn More
Trajan Scientific and Medical™ Replacement Needles for CTC Analytics Plunger-in-Needle Syringes
A range of quality needles to fit a SGE syringe in a range of lengths and tip styles
Brand:  Trajan Scientific and Medical™ 034905
Additional Details : Weight : 0.01000kg
Product Code. 15268583
  • 71.20 EUR / Each
Estimated Shipment: 11-07-2024
to see stock.
Description
Description
  • Needle length is the exposed length of needle
  • Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
Replacement Needle
0.22 mm
2 μL
23 G
0.47 mm
CTC Analytics Plunger-in-Needle Syringes