Learn More
Trajan Scientific and Medical™ Replacement Needles for 1-2.5mL Syringes
Range of quality needles to fit a SGE syringe in a range of lengths and tip styles.
Brand:  Trajan Scientific and Medical™ 039115
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15288603
  • 51.95 EUR / Pack of 2
In Stock 1
Add to basket
Description
Description
  • Needle length is the exposed length of needle
  • Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
22 G
0.71mm
50mm
0.37mm
1-2.5mL
Bevel Tipped Needle