Learn More
Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths
Reduce operating noise and potential escape of aerosols. Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths are supplied as standard with the bath.
Brand: Grant Instruments™ XAL5
Additional Details : Weight : 1.00000kg
Description
- ABS construction
- For use with XUB and XUBA Series Ultrasonic Baths
Specifications
XAB5 Basket for Ultrasonic Bath | |
ABS |