Learn More
Thermo Scientific™ Repair Kits for Thermo Scientific™ Plunger-in-Needle Syringes
Extend the life and performance, and reduce the cost per use of your syringes with the Thermo Scientific™ Repair Kit for Thermo Scientific Plunger-in-Needle Syringes.
Brand: Thermo Scientific™ 365RK341
Additional Details : Weight : 0.05000kg
Description
Thermo Scientific™ Repair Kits for Thermo Scientific Plunger-in-Needle Syringes extend the life and performance and reduce cost per use of syringes.
- Repair kits for plunger in needle syringes contain both the plunger and needle, which must be changed together
Includes: Plunger, 1; Needle, 1
Specifications
For PIN syringe 365B0531 | |
2.7559 in. | |
1 μL | |
PIN Syringe 365B0531 |
23 G | |
Syringe Repair Kit | |
70 mm | |
Plunger-In |