Learn More
Thermo Scientific™ Remel™ Letheen Broth (Dehydrated)
The Remel Letheen Broth (Dehydrated) is used for testing quaternary ammonium compounds for antimicrobial activity.
195.00 EUR
Specifications
Certifications/Compliance | Industrial Reference: AOAC. |
---|---|
Description | Letheen Broth |
Form | Powder |
Product Type | Dehydrated Culture Media |
Description
Remel Letheen Broth (Dehydrated) is used for testing quaternary ammonium compounds for antimicrobial activity.
Specifications
Industrial Reference: AOAC. | |
Letheen Broth | |
Powder | |
Dehydrated Culture Media |
REF R07700, REF R112420, REF R112423, and REF R117259 not intended for IVD use.