Learn More
Thermo Scientific™ Remel™ Letheen Broth (Dehydrated)
The Remel Letheen Broth (Dehydrated) is used for testing quaternary ammonium compounds for antimicrobial activity.
195.00 EUR
Specifications
Certifications/Compliance Industrial Reference: AOAC.
Description Letheen Broth
Form Powder
Product Type Dehydrated Culture Media
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13215949
View Documents Promotion Details
Thermo Scientific™
R453712
500 g
195.00 EUR
500g
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
Description
Remel Letheen Broth (Dehydrated) is used for testing quaternary ammonium compounds for antimicrobial activity.
Specifications
Specifications
Industrial Reference: AOAC.
Letheen Broth
Powder
Dehydrated Culture Media
Documents
Product Certifications
REF R07700, REF R112420, REF R112423, and REF R117259 not intended for IVD use.