Learn More
Thermo Scientific™ Remel™ Colorimeter Standard Set, Vitek™
Thermo Scientific™ Remel Colorimeter Standard Kit contains latex equivalence turbidity standards for use with Vitek colorimeter.
Brand: Thermo Scientific™ R20343
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ Remel Colorimeter Standard Kit contains latex equivalence turbidity standards for use with Vitek colorimeter.
Contains one each of 0.5, 1.0, 2.0, and 3.0 standards.Specifications
Colorimeter Standard Set for use with Vitek™ | |
one each of 0.5, 1.0, 2.0, and 3.0 standards |