Learn More
Fisherbrand™ Reinforced Silicone Tubing
Providing excellent crushing resistance and tear strength across a wide range of applications. Fisherbrand™ Reinforced Silicone Tubing is oderless and non-toxic.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11753974
  • 419.00 EUR / Each
Estimated Shipment: 05-08-2024
Description
  • Autoclavable
  • Glossy in appearance
  • Translucent and flexible
  • FDA (food) compliant
  • Applications include pharmaceutical, high pressure and temperature work, biotechnology and aerospace
Specifications
Specifications
FDA, BFR and WRAS approved
5 mm
25 m
40 bar
Autoclavable
11.4 mm
3.2 mm