Learn More
Brady™ Reinforced Polypropylene Mandatory Signs
Brand:  Brady™ 252139
Additional Details : Weight : 0.05300kg
 View more versions of this product
Product Code. 10623965
  • 13.55 EUR / Each
Estimated Shipment: 05-06-2024
Specifications
Polypropylene
Reinforced Polypropylene
120°C
Round
Mandatory Sign
1 Ea.
200 mm
0.369 mm
Use Protective Hood
White on Blue