missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zebrafish Metallothionein-1 Protein
A DNA sequence encoding the Danio rerio (Zebrafish) (Brachydanio rerio) Metallothionein-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7223-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_571150.1 | |
Metallothionein-1 Protein | |
Research Use Only | |
mt | |
Recombinant Protein | |
E. coli | |
MDPCECAKTG ACNCGATCKC TNCQCTTCKK SCCSCCPSGC SKCASGCVCK GNSCGTSCCQ | |
Greater than 90% as determined by SDS-PAGE. |
30282 | |
500 μg | |
Not determined. | |
Full Length | |
Zebrafish | |
His-SUMO | |
Tris-based buffer, 50% glycerol |