missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep TGF-alpha Protein
A DNA sequence encoding the Ovis aries (Sheep) TGF-alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6777-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
TGF-alpha Protein | |
Research Use Only | |
TGFA | |
Recombinant Protein | |
E. coli | |
ENSTSALSDP PVAAAVVSHF NDCPDSHTQF CFHGTCRFLL QEEKPACVCH SGYVGARCEH ADLLAVVAAS QKKQ | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Extracellular Domain | |
Sheep | |
GST | |
Tris-based buffer, 50% glycerol |