missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep IL-1 beta / IL1B Protein
A DNA sequence encoding the Ovis aries (Sheep) IL-1 beta / IL1B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6215-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001009465.2 | |
IL-1 beta / IL1B Protein | |
Research Use Only | |
IL1B | |
Recombinant Protein | |
E. coli | |
AAVQSVKCKL QDREQKSLVL DSPCVLKALH LPSQEMSREV VFCMSFVQGE ERDNKIPVAL GIRDKNLYLS CVKKGDTPTL QLEEVDPKVY PKRNMEKRFV FYKTEIKNTV EFESVLYPNW YISTSQIEEK PVFLGRFRGG QDITDFRMET LSP | |
Greater than 90% as determined by SDS-PAGE. |
443539 | |
10 μg | |
Not determined. | |
Full Length | |
Sheep | |
His | |
Tris-based buffer, 50% glycerol |