missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Sheep IL-1 beta / IL1B Protein

A DNA sequence encoding the Ovis aries (Sheep) IL-1 beta / IL1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6215-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031640

  • 2985.00 EUR / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001009465.2
IL-1 beta / IL1B Protein
Research Use Only
IL1B
Recombinant Protein
E. coli
AAVQSVKCKL QDREQKSLVL DSPCVLKALH LPSQEMSREV VFCMSFVQGE ERDNKIPVAL GIRDKNLYLS CVKKGDTPTL QLEEVDPKVY PKRNMEKRFV FYKTEIKNTV EFESVLYPNW YISTSQIEEK PVFLGRFRGG QDITDFRMET LSP
Greater than 90% as determined by SDS-PAGE.
443539
1 mg
Not determined.
Full Length
Sheep
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers