missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Ubiquitin-like protein SMT3 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Ubiquitin-like protein SMT3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7546-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_010798.1 | |
S. cerevisiae Ubiquitin-like protein SMT3 Protein | |
Research Use Only | |
SMT3 | |
Recombinant Protein | |
E. coli | |
SDSEVNQEAK PEVKPEVKPE THINLKVSDG SSEIFFKIKK TTPLRRLMEA FAKRQGKEMD SLRFLYDGIR IQADQTPEDL DMEDNDIIEA HREQIGG | |
Greater than 90% as determined by SDS-PAGE. |
852122 | |
1 mg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |