missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae SPT15 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) SPT15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6764-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035670

  • 2325.00 EUR / 500µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

S. cerevisiae SPT15 Protein
Research Use Only
SPT15
Recombinant Protein
Yeast
ADEERLKEFK EANKIVFDPN TRQVWENQNR DGTKPATTFQ SEEDIKRAAP ESEKDTSATS GIVPTLQNIV ATVTLGCRLD LKTVALHARN AEYNPKRFAA VIMRIREPKT TALIFASGKM VVTGAKSEDD SKLASRKYAR IIQKIGFAAK FTDFKIQNIV GSCDVKFPIR LEGLAFSHGT FSSYEPELFP GLIYRMVKPK IVLLIFVSGK IVLTGAKQRE EIYQAFEAIY PVLSEFRKM
Greater than 90% as determined by SDS-PAGE.
500 μg
Not determined.
Full Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers