missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Serine--tRNA ligase, cytoplasmic Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Serine--tRNA ligase, cytoplasmic, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7297-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068070

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_010306.1
S. cerevisiae Serine--tRNA ligase, cytoplasmic Protein
Research Use Only
SES1
Recombinant Protein
Yeast
MLDINQFIED KGGNPELIRQ SQKARNASVE IVDEIISDYK DWVKTRFELD ELNKKFNKLQ KDIGLKFKNK EDASGLLAEK EKLTQQKKEL TEKEQQEDKD LKKKVFQVGN IVHPSVVVSN DEENNELVRT WKPEDLEAVG PIASVTGKPA SLSHHEILLR LDGYDPDRGV KICGHRGYFF RNYGVFLNQA LINYGLQFLA AKGYIPLQAP VMMNKELMSK TAQLSEFDEE LYKVIDGEDE KYLIATSEQP ISAYHSGEWF EKPQEQLPIH YVGYSSCFRR EAGSHGKDAW GVFRVHAFEK IEQFVITEPE KSWEEFEKMI SYSEEFYKSL KLPYRIVGIV SGELNNAAAK KYDLEAWFPY QKEYKELVSC SNCTDYQSRN LEIRCGIKKM GDREKKYVHC LNSTLAATQR ALCCILENYQ TEDGLVVPEV LRKYIPGEPE FLPFVNELPK NSTSSKDKKK KN
Greater than 90% as determined by SDS-PAGE.
851587
50 μg
Not determined.
Full Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers