missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Serine--tRNA ligase, cytoplasmic Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Serine--tRNA ligase, cytoplasmic, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7297-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_010306.1 | |
S. cerevisiae Serine--tRNA ligase, cytoplasmic Protein | |
Research Use Only | |
SES1 | |
Recombinant Protein | |
Yeast | |
MLDINQFIED KGGNPELIRQ SQKARNASVE IVDEIISDYK DWVKTRFELD ELNKKFNKLQ KDIGLKFKNK EDASGLLAEK EKLTQQKKEL TEKEQQEDKD LKKKVFQVGN IVHPSVVVSN DEENNELVRT WKPEDLEAVG PIASVTGKPA SLSHHEILLR LDGYDPDRGV KICGHRGYFF RNYGVFLNQA LINYGLQFLA AKGYIPLQAP VMMNKELMSK TAQLSEFDEE LYKVIDGEDE KYLIATSEQP ISAYHSGEWF EKPQEQLPIH YVGYSSCFRR EAGSHGKDAW GVFRVHAFEK IEQFVITEPE KSWEEFEKMI SYSEEFYKSL KLPYRIVGIV SGELNNAAAK KYDLEAWFPY QKEYKELVSC SNCTDYQSRN LEIRCGIKKM GDREKKYVHC LNSTLAATQR ALCCILENYQ TEDGLVVPEV LRKYIPGEPE FLPFVNELPK NSTSSKDKKK KN | |
Greater than 90% as determined by SDS-PAGE. |
851587 | |
10 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |