missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Pyruvate carboxylase 1 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Pyruvate carboxylase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7045-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007160

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_011453.1
S. cerevisiae Pyruvate carboxylase 1 Protein
Research Use Only
PYC1
Recombinant Protein
E. coli
EKNKILVANR GEIPIRIFRT AHELSMQTVA IYSHEDRLST HKQKADEAYV IGEVGQYTPV GAYLAIDEII SIAQKHQVDF IHPGYGFLSE NSEFADKVVK AGITWIGPPA EVIDSVGDKV SARNLAAKAN VPTVPGTPGP IETVEEALDF VNEYGYPVII KAAFGGGGRG MRVVREGDDV ADAFQRATSE ARTAFGNGTC FVERFLDKPK HIEVQLLADN HGNVVHLFER DCSVQRRHQK VVEVAPAKTL PREVRDAILT DAVKLAKECG YRNAGTAEFL VDNQNRHYFI EINPRIQVEH TITEEITGID IVAAQIQIAA GASLPQLGLF QDKITTRGFA IQCRITTEDP AKNFQPDTGR IEVYRSAGGN GVRLDGGNAY AGTIISPHYD SMLVKCSCSG STYEIVRRKM IRALIEFRIR GVKTNIPFLL TLLTNPVFIE GTYWTTFIDD TPQ
Greater than 90% as determined by SDS-PAGE.
852818
500 μg
Not determined.
Partial (see sequence information for more details).
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers