missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Neutral trehalase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Neutral trehalase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8922-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_010284.1 | |
S. cerevisiae Neutral trehalase Protein | |
Research Use Only | |
NTH1 | |
Recombinant Protein | |
E. coli | |
YMWDDESGFF FDYNTKIKHR TSYESATTFW ALWAGLATKE QAQKMVEKAL PKLEMLGGLA ACTERSRGPI SISRPIRQWD YPFGWAPHQI LAWEGLRSYG YLTVTNRLAY RWLFMMTKAF VDYNGIVVEK YDVTRGTDPH RVEAEYGNQG ADFKGAATEG FGWVNASYIL GLKYMNSHAR RALGACIPPI SFFSSLRPQE RNLYGL | |
Greater than 90% as determined by SDS-PAGE. |
851564 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |