missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Neutral trehalase Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Neutral trehalase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8922-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047191

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_010284.1
S. cerevisiae Neutral trehalase Protein
Research Use Only
NTH1
Recombinant Protein
E. coli
YMWDDESGFF FDYNTKIKHR TSYESATTFW ALWAGLATKE QAQKMVEKAL PKLEMLGGLA ACTERSRGPI SISRPIRQWD YPFGWAPHQI LAWEGLRSYG YLTVTNRLAY RWLFMMTKAF VDYNGIVVEK YDVTRGTDPH RVEAEYGNQG ADFKGAATEG FGWVNASYIL GLKYMNSHAR RALGACIPPI SFFSSLRPQE RNLYGL
Greater than 90% as determined by SDS-PAGE.
851564
200 μg
Not determined.
Partial (see sequence information for more details).
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers