missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae GTP-binding nuclear protein GSP1 / CNR1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) GTP-binding nuclear protein GSP1/CNR1, was expressed in the hosts and tags indicated.
352.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_013396.1 |
---|---|
Gene ID (Entrez) | 851000 |
Name | S. cerevisiae GTP-binding nuclear protein GSP1 / CNR1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16007540
|
enQuireBio™
QP7163-YE-100UG |
100 μg |
965.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047540
|
enQuireBio™
QP7163-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017540
|
enQuireBio™
QP7163-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037540
|
enQuireBio™
QP7163-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057540
|
enQuireBio™
QP7163-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027540
|
enQuireBio™
QP7163-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_013396.1 | |
S. cerevisiae GTP-binding nuclear protein GSP1 / CNR1 Protein | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |
851000 | |
Research Use Only | |
GSP1 | |
Recombinant Protein | |
Yeast | |
SAPAANGEVP TFKLVLVGDG GTGKTTFVKR HLTGEFEKKY IATIGVEVHP LSFYTNFGEI KFDVWDTAGQ EKFGGLRDGY YINAQCAIIM FDVTSRITYK NVPNWHRDLV RVCENIPIVL CGNKVDVKER KVKAKTITFH RKKNLQYYDI SAKSNYNFEK PFLWLARKLA GNPQLEFVAS PALAPPEVQV DEQLMQQYQQ EMEQATALPL PDEDDADL | |
Greater than 90% as determined by SDS-PAGE. |