missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Exopolyphosphatase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Exopolyphosphatase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7169-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_012071.1 | |
S. cerevisiae Exopolyphosphatase Protein | |
Research Use Only | |
PPX1 | |
Recombinant Protein | |
E. coli | |
MSPLRKTVPE FLAHLKSLPI SKIASNDVLT ICVGNESADM DSIASAITYS YCQYIYNEGT YSEEKKKGSF IVPIIDIPRE DLSLRRDVMY VLEKLKIKEE ELFFIEDLKS LKQNVSQGTE LNSYLVDNND TPKNLKNYID NVVGIIDHHF DLQKHLDAEP RIVKVSGSCS SLVFNYWYEK LQGDREVVMN IAPLLMGAIL IDTSNMRRKV EESDKLAIER CQAVLSGAVN EVSAQGLEDS SEFYKEIKSR KNDIKGFSVS DILKKDYKQF NFQGKGHKGL EIGLSSIVKR MSWLFNEHGG EADFVNQCRR FQAERGLDVL VLLTSWRKAG DSHRELVILG DSNVVRELIE RVSDKLQLQL FGGNLDGGVA MFKQLNVEAT RKQVVPYLEE AYSNLEE | |
Greater than 90% as determined by SDS-PAGE. |
856608 | |
10 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |