missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Dipeptidyl aminopeptidase A Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Dipeptidyl aminopeptidase A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7196-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_014862.3 | |
S. cerevisiae Dipeptidyl aminopeptidase A Protein | |
Research Use Only | |
STE13 | |
Recombinant Protein | |
E. coli | |
MSASTHSHKR KNSHLFPQRK SSNSSMDKPF FPNNDSVANT DPQSNENGHT INEIRPTEAT IDVTDVPQTP FLQEQYSMRP RRESFQFNDI ENQHHTHSFF SVNKFNRRWG EWSLPEKRS | |
Greater than 90% as determined by SDS-PAGE. |
854394 | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |