missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Cystathionine beta-synthase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Cystathionine beta-synthase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8802-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_011671.3 | |
S. cerevisiae Cystathionine beta-synthase Protein | |
Research Use Only | |
CYS4 | |
Recombinant Protein | |
E. coli | |
MTKSEQQADS RHNVIDLVGN TPLIALKKLP KALGIKPQIY AKLELYNPGG SIKDRIAKSM VEEAEASGRI HPSRSTLIEP TSGNTGIGLA LIGAIKGYRT IITLPEKMSN EKVSVLKALG AEIIRTPTAA AWDSPESHIG VAKKLEKEIP GAVILDQYNN MMNPEAHYFG TGREIQRQLE DLNLFDNLRA VVAGAGTGGT ISGISKYLKE QNDKIQIVGA DPFGSILAQP ENLNKTDITD YKVEGIGYDF VPQVLDRKLI DVWYKTDDKP SFKYARQLIS NEGVLVGGSS GSAFTAVVKY CEDHPELTED DVIVAIFPDS IRSYLTKFVD DEWLKKNNLW DDDVLARFDS SKLEASTTKY ADVFGNATVK DLHLKPVVSV KETAKVTDVI KILKDNGFDQ LPVLTEDGKL SGLVTLSELL RKLSINNSNN DNTIKGKYLD FKKLNNFNDV SSYNENKSGK KKFIKFDENS KLSDLNRFFE KNSSAVITDG LKPIHIVTKM DLLSYLA | |
Greater than 90% as determined by SDS-PAGE. |
853059 | |
500 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |