missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Adenosylhomocysteinase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Adenosylhomocysteinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8805-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_010961.3 | |
S. cerevisiae Adenosylhomocysteinase Protein | |
Research Use Only | |
SAH1 | |
Recombinant Protein | |
E. coli | |
MSAPAQNYKI ADISLAAFGR KEIELAEHEM PGLMAIRKAY GDVQPLKGAR IAGCLHMTIQ TAVLIETLVA LGAEVTWSSC NIYSTQDHAA AAIAASGVPV FAWKGETEEE YLWCIEQQLF AFKDNKKLNL ILDDGGDLTT LVHEKHPEML EDCFGLSEET TTGVHHLYRM VKEGKLKVPA INVNDSVTKS KFDNLYGCRE SLVDGIKRAT DVMLAGKVAV VAGYGDVGKG CAAALRGMGA RVLVTEIDPI NALQAAMEGY QVVTMEDASH IGQVFVTTTG CRDIINGEHF INMPEDAIVC NIGHFDIEID VAWLKANAKE CINIKPQVDR YLLSSGRHVI LLANGRLVNL GCATGHSSFV MSCSFSNQVL AQIALFKSND KSFREKHIEF QKTGPFEVGV HVLPKILDEA VAKFHLGNLG VRLTKLSKVQ SEYLGIPEEG PFKADHYR | |
Greater than 90% as determined by SDS-PAGE. |
856766 | |
100 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |