missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Adenosylhomocysteinase Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Adenosylhomocysteinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8805-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036571

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_010961.3
S. cerevisiae Adenosylhomocysteinase Protein
Research Use Only
SAH1
Recombinant Protein
E. coli
MSAPAQNYKI ADISLAAFGR KEIELAEHEM PGLMAIRKAY GDVQPLKGAR IAGCLHMTIQ TAVLIETLVA LGAEVTWSSC NIYSTQDHAA AAIAASGVPV FAWKGETEEE YLWCIEQQLF AFKDNKKLNL ILDDGGDLTT LVHEKHPEML EDCFGLSEET TTGVHHLYRM VKEGKLKVPA INVNDSVTKS KFDNLYGCRE SLVDGIKRAT DVMLAGKVAV VAGYGDVGKG CAAALRGMGA RVLVTEIDPI NALQAAMEGY QVVTMEDASH IGQVFVTTTG CRDIINGEHF INMPEDAIVC NIGHFDIEID VAWLKANAKE CINIKPQVDR YLLSSGRHVI LLANGRLVNL GCATGHSSFV MSCSFSNQVL AQIALFKSND KSFREKHIEF QKTGPFEVGV HVLPKILDEA VAKFHLGNLG VRLTKLSKVQ SEYLGIPEEG PFKADHYR
Greater than 90% as determined by SDS-PAGE.
856766
200 μg
Not determined.
Full Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers