missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Acyl-CoA-binding Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Acyl-CoA-binding protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9099-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_011551.3 | |
S. cerevisiae Acyl-CoA-binding Protein | |
Research Use Only | |
ACB1 | |
Recombinant Protein | |
Yeast | |
MVSQLFEEKA KAVNELPTKP STDELLELYA LYKQATVGDN DKEKPGIFNM KDRYKWEAWE NLKGKSQEDA EKEYIALVDQ LIAKYSS | |
Greater than 90% as determined by SDS-PAGE. |
852925 | |
100 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |