missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Transposase for transposon Tn552 Protein

A DNA sequence encoding the Staphylococcus aureus Transposase for transposon Tn552, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7085-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007340

  • 555.00 EUR / 50µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_000790705.1
S. aureus Transposase for transposon Tn552 Protein
Research Use Only
Tn552
Recombinant Protein
E. coli
MKNKEKYLTN FSEAKRKEAT QKYNIIKPFI LGKQSLSSIS KSKGIALSTL YRWNKLYKEQ GLTGLIHNTR VDKGEHKLKQ NIIDEIKRLA LKNKRNSIAT IHRKIANYCI ENNFYKPSYK QVYSIIKAMP KSVIDFSHQG EKYYQNKYDL IQIRESSRPN EIWQADHTLL DIYILDQKGN INRPWLTIIM DDYSRAIAGY FISFDAPNAQ NTALTLHQAI WNKNNTNWPV CGIPEKFYTD HGSDFTSHHM EQVAIDLKIN LMFSKVGVPR GRGKIERFFQ TVNQTFLEQL PGYINNNDTS SDLIDFQNFE EKLRYFLIED YNQKEHSAIQ STPINRWNSN HFFPNMPSSL EQLDLLLLEI PKSRKIHSDG IHFQGFRYSN TNLTAYVGEY VLIRYNPNDM AEIRVFYRDE FLCTAISPDL ADYSIDIKEI QHARSQRRKH LKQNIASPST TDLIKEEKSY GYSPQETTKN VKKLKRYRND
Greater than 90% as determined by SDS-PAGE.
13875815
50 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers