missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Transposase for transposon Tn552 Protein
A DNA sequence encoding the Staphylococcus aureus Transposase for transposon Tn552, was expressed in the hosts and tags indicated.
385.00 EUR - 3285.00 EUR
Specifications
Accession Number | WP_000790705.1 |
---|---|
Gene ID (Entrez) | 13875815 |
Name | S. aureus Transposase for transposon Tn552 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16067330
|
enQuireBio™
QP7085-10UG |
10 μg |
385.00 EUR
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007340
|
enQuireBio™
QP7085-50UG |
50 μg |
555.00 EUR
50µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057330
|
enQuireBio™
QP7085-100UG |
100 μg |
940.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087330
|
enQuireBio™
QP7085-200UG |
200 μg |
1550.00 EUR
200µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097330
|
enQuireBio™
QP7085-500UG |
500 μg |
2070.00 EUR
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077330
|
enQuireBio™
QP7085-1MG |
1 mg |
3285.00 EUR
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
WP_000790705.1 | |
S. aureus Transposase for transposon Tn552 Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
13875815 | |
Research Use Only | |
Tn552 | |
Recombinant Protein | |
E. coli | |
MKNKEKYLTN FSEAKRKEAT QKYNIIKPFI LGKQSLSSIS KSKGIALSTL YRWNKLYKEQ GLTGLIHNTR VDKGEHKLKQ NIIDEIKRLA LKNKRNSIAT IHRKIANYCI ENNFYKPSYK QVYSIIKAMP KSVIDFSHQG EKYYQNKYDL IQIRESSRPN EIWQADHTLL DIYILDQKGN INRPWLTIIM DDYSRAIAGY FISFDAPNAQ NTALTLHQAI WNKNNTNWPV CGIPEKFYTD HGSDFTSHHM EQVAIDLKIN LMFSKVGVPR GRGKIERFFQ TVNQTFLEQL PGYINNNDTS SDLIDFQNFE EKLRYFLIED YNQKEHSAIQ STPINRWNSN HFFPNMPSSL EQLDLLLLEI PKSRKIHSDG IHFQGFRYSN TNLTAYVGEY VLIRYNPNDM AEIRVFYRDE FLCTAISPDL ADYSIDIKEI QHARSQRRKH LKQNIASPST TDLIKEEKSY GYSPQETTKN VKKLKRYRND | |
Greater than 90% as determined by SDS-PAGE. |