missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Staphopain B Protein

A DNA sequence encoding the Staphylococcus aureus Staphopain B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8022-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042211

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_001088793.1
500 μg
Not determined.
Full Length
S. aureus
GST
Tris-based buffer, 50% glycerol
S. aureus Staphopain B Protein
Research Use Only
sspB
Recombinant Protein
E. coli
DQVQYENTLK NFKIREQQFD NSWCAGFSMA ALLNATKNTD TYNAHDIMRT LYPEVSEQDL PNCATFPNQM IEYGKSQGRD IHYQEGVPSY NQVDQLTKDN VGIMILAQSV SQNPNDPHLG HALAVVGNAK INDQEKLIYW NPWDTELSIQ DADSSLLHLS FNRDYNWYGS MIGY
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers