missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus fabF Protein

A DNA sequence encoding the Staphylococcus aureus (strain COL) fabF, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7667-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040081

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_000081240.1
S. aureus fabF Protein
Research Use Only
fabF
Recombinant Protein
E. coli
MSQNKRVVIT GMGALSPIGN DVKTTWENAL KGVNGIDKIT RIDTEPYSVH LAGELKNFNI EDHIDKKEAR RMDRFTQYAI VAAREAVKDA QLDINENTAD RIGVWIGSGI GGMETFEIAH KQLMDKGPRR VSPFFVPMLI PDMATGQVSI DLGAKGPNGA TVTACATGTN SIGEAFKIVQ RGDADAMITG GTEAPITHMA IAGFSASRAL STNDDIETAC RPFQEGRDGF VMGEGAGILV IESLESAQAR GANIYAEIVG YGTTGDAYHI TAPAPEGEGG SRAMQAAMDD AGIEPKDVQY LNAHGTSTPV GDLNEVKAIK NTFGEAAKHL KVSSTKSMTG HLLGATGGIE AIFSALSIKD SKVAPTIHAV TPDPECDLDI VPNEAQDLDI TYAMSNSLGF GGHNAVLVFK KFEA
Greater than 90% as determined by SDS-PAGE.
31213734
200 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers