missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus esxA Protein

A DNA sequence encoding the Staphylococcus aureus (strain MSSA476) esxA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7776-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020721

  • 860.00 EUR / 100µg
Estimated Shipment: 02-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_001240826.1
S. aureus esxA Protein
100 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
31213083
Greater than 90% as determined by SDS-PAGE.
Research Use Only
esxA
Recombinant Protein
E. coli
MAMIKMSPEE IRAKSQSYGQ GSDQIRQILS DLTRAQGEIA ANWEGQAFSR FEEQFQQLSP KVEKFAQLLE EIKQQLNSTA DAVQEQDQQL SNNFGLQ
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers