missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type H Protein
A DNA sequence encoding the Staphylococcus aureus Enterotoxin type H, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7312-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000608674.1 | |
500 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus Enterotoxin type H Protein | |
Research Use Only | |
entH | |
Recombinant Protein | |
E. coli | |
EDLHDKSELT DLALANAYGQ YNHPFIKENI KSDEISGEKD LIFRNQGDSG NDLRVKFATA DLAQKFKNKN VDIYGASFYY KCEKISENIS ECLYGGTTLN SEKLAQERVI GANVWVDGIQ KETELIRTNK KNVTLQELDI KIRKILSDKY KIYYKDSEIS KGLIEFDMKT PRDYSFDIYD LKGENDYEID KIYEDNKTLK SDDISHIDVN LYTKKKV | |
Greater than 90% as determined by SDS-PAGE. |