missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type E Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Pyruvate carboxylase 1, was expressed in the hosts and tags indicated.
352.00 EUR - 3575.00 EUR
Specifications
Name | S. aureus Enterotoxin type E Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Type | Recombinant Protein |
Species | Yeast |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16007180
|
enQuireBio™
QP7046-YE-100UG |
100 μg |
965.00 EUR
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027180
|
enQuireBio™
QP7046-YE-1MG |
1 mg |
3575.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16017180
|
enQuireBio™
QP7046-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037180
|
enQuireBio™
QP7046-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047180
|
enQuireBio™
QP7046-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057180
|
enQuireBio™
QP7046-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
S. aureus Enterotoxin type E Protein | |
Not determined. | |
Yeast | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
Recombinant Protein | |
His | |
Greater than 90% as determined by SDS-PAGE. |