missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type A Protein
A DNA sequence encoding the Staphylococcus aureus Enterotoxin type A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8860-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000750412.1 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |
S. aureus Enterotoxin type A Protein | |
Research Use Only | |
entA | |
Recombinant Protein | |
E. coli | |
SEKSEEINEK DLRKKSELQG TALGNLKQIY YYNEKAKTEN KESHDQFLQH TILFKGFFTD HSWYNDLLVD FDSKDIVDKY KGKKVDLYGA YYGYQCAGGT PNKTACMYGG VTLHDNNRLT EEKKVPINLW LDGKQNTVPL ETVKTNKKNV TVQELDLQAR RYLQEKYNLY NSDVFDGKVQ RGLIVFHTST EPSVNYDLFG AQGQYSNTLL RIYRDNKTIN SENMHIDIY | |
Greater than 90% as determined by SDS-PAGE. |