missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type A Protein
A DNA sequence encoding the Staphylococcus aureus Enterotoxin type A, was expressed in the hosts and tags indicated.
309.00 EUR - 2645.00 EUR
Specifications
Accession Number | WP_000750412.1 |
---|---|
Name | S. aureus Enterotoxin type A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | entA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006881
|
enQuireBio™
QP8860-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046881
|
enQuireBio™
QP8860-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096871
|
enQuireBio™
QP8860-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026881
|
enQuireBio™
QP8860-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036881
|
enQuireBio™
QP8860-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016881
|
enQuireBio™
QP8860-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000750412.1 | |
Research Use Only | |
entA | |
Recombinant Protein | |
E. coli | |
SEKSEEINEK DLRKKSELQG TALGNLKQIY YYNEKAKTEN KESHDQFLQH TILFKGFFTD HSWYNDLLVD FDSKDIVDKY KGKKVDLYGA YYGYQCAGGT PNKTACMYGG VTLHDNNRLT EEKKVPINLW LDGKQNTVPL ETVKTNKKNV TVQELDLQAR RYLQEKYNLY NSDVFDGKVQ RGLIVFHTST EPSVNYDLFG AQGQYSNTLL RIYRDNKTIN SENMHIDIY | |
Greater than 90% as determined by SDS-PAGE. |
S. aureus Enterotoxin type A Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |