missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enolase Protein
A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Enolase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7255-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
WP_001121760.1 | |
S. aureus Enolase Protein | |
Research Use Only | |
eno | |
Recombinant Protein | |
E. coli | |
MPIITDVYAR EVLDSRGNPT VEVEVLTESG AFGRALVPSG ASTGEHEAVE LRDGDKSRYL GKGVTKAVEN VNEIIAPEII EGEFSVLDQV SIDKMMIALD GTPNKGKLGA NAILGVSIAV ARAAADLLGQ PLYKYLGGFN GKQLPVPMMN IVNGGSHSDA PIAFQEFMIL PVGATTFKES LRWGTEIFHN LKSILSKRGL ETAVGDEGGF APKFEGTEDA VETIIQAIEA AGYKPGEEVF LGFDCASSEF YENGVYDYSK FEGEHGAKRT AAEQVDYLEQ LVDKYPIITI EDGMDENDWD GWKQLTERIG DRVQLVGDDL FVTNTEILAK GIENGIGNSI LIKVNQIGTL TETFDAIEMA QKAGYTAVVS HRSGETEDTT IADIAVATNA GQIKTGSLSR TDRIAKYNQL LRIEDELFET AKYDGIKSFY NLDK | |
Greater than 90% as determined by SDS-PAGE. |
31213594 | |
10 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |