missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Vesicle-fusing ATPase Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Vesicle-fusing ATPase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8832-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056721

  • 2985.00 EUR / 1mg
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_068516.1
Vesicle-fusing ATPase Protein
Research Use Only
Nsf
Recombinant Protein
E. coli
QAARCPTDEL SLSNCAVVNE KDYQSGQHVM VRTSPNHKYI FTLRTHPSVV PGCIAFSLPQ RKWAGLSIGQ DIEVALYSFD KAKQCIGTMT IEIDFLQKKN IDSNPYDTDK MAAEFIQQFN HQAFSVGQQL VFSFNDKLFG LLVKDIEAMD PSILKGEPAS GKRQKIEVGL VVGNSQVAFE KAENSSLNLI GKAKTKENRQ SII
Greater than 90% as determined by SDS-PAGE.
60355
1 mg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers