missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Vasopressin V1b receptor Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Vasopressin V1b receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8844-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
Vasopressin V1b receptor Protein | |
Research Use Only | |
Avpr1b | |
Recombinant Protein | |
E. coli | |
NSRLLPRSLS HHACCTGSKP QVHRQLSTSS LTSRRTTLLT HACGSPTLRL SLNLSLRAKP RPAGSLKDLE QVDGEATMET SIF | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |