missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Vasopressin V1a receptor Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Vasopressin V1a receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8843-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_444178.2 | |
Vasopressin V1a receptor Protein | |
Research Use Only | |
Avpr1a | |
Recombinant Protein | |
E. coli | |
SQDRSVGNSS PWWPLTTEGS NGSQEAARLG EGDSPLGDVR NEELAK | |
Greater than 90% as determined by SDS-PAGE. |
25107 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |