missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Trophoblast glycoProtein
A DNA sequence encoding the Rattus norvegicus (Rat) Trophoblast glycoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6824-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_113995.1 | |
Trophoblast glycoProtein | |
Research Use Only | |
Tpbg | |
Recombinant Protein | |
E. coli | |
SAPSSSLPSS STSPAAFLAS GSAQPPPAER CPAACECSEA ARTVKCVNRN LLEVPADLPP YVRNLFLTGN QMTVLPAGAF ARQPPLADLA VLNLSGNHLK EVGAGAFEHL PGLRRLDLSH NPLTNLSAFT FAGSNVSVST PSPLLELILN HIVPPEDQRQ NGSFEGMVAF EGMVAAALRS GLALRGLHHL ELASNHFLYL PRDLLDQLPS LKHLDLRNNS LVSLTYASFR NLTHLESLHL EDNALKVLHN STLAEWQGLA HVRVFLDNNP WVCDCYMADM VSWLKETEVV PDKARLTCAF PEKMRNRGLL DLTSSDLDCD ATLPQSLQTS | |
Greater than 90% as determined by SDS-PAGE. |
83684 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |