missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Thyroxine 5-deiodinase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Thyroxine 5-deiodinase, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_058906.3 |
---|---|
Gene ID (Entrez) | 29475 |
Name | Thyroxine 5-deiodinase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16077770
|
enQuireBio™
QP7221-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097770
|
enQuireBio™
QP7221-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087770
|
enQuireBio™
QP7221-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007780
|
enQuireBio™
QP7221-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017780
|
enQuireBio™
QP7221-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027780
|
enQuireBio™
QP7221-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_058906.3 | |
Thyroxine 5-deiodinase Protein | |
Not determined. | |
Extracellular Domain | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
29475 | |
Research Use Only | |
Dio3 | |
Recombinant Protein | |
E. coli | |
DFLCIRKHFL RRRHPDHPEP EVELNSEGEE MPPDDPPICV SDDNRLCTLA SLKAVWHGQK LDFFKQAHEG GPAPNSEVVR PDGFQSQRIL DYAQGTRPLV LNFGSCTUPP FMARMSAFQR LVTKYQRDVD FLIIYIEEAH PSDGWVTTDS PYVIPQHRSL EDRVSAARVL QQGAPGCALV LDTMANSSSS AYGAYFERLY VIQSGTIMYQ GGRGPDGYQV SELRTWLERY DEQLHGTRPR RL | |
Greater than 90% as determined by SDS-PAGE. |