missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat TGF-beta 1 / TGFB1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6780-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_067589.1 | |
TGF-beta 1 / TGFB1 Protein | |
Research Use Only | |
TGFB1 | |
Recombinant Protein | |
E. coli | |
LSTCKTIDME LVKRKRIEAI RGQILSKLRL ASPPSQGEVP PGPLPEAVLA LYNSTRDRVA GESADPEPEP EADYYAKEVT RVLMVDRNNA IYDKTKDITH SIYMFFNTSD IREAVPEPPL LSRAELRLQR FKSTVEQHVE LYQKYSNNSW RYLGNRLLTP TDTPEWLSFD VTGVVRQWLN QGDGIQGFRF SAHCSCDSKD NVLHVEINGI SPKRRGDLGT IHDMNRPFLL LMATPLERAQ HLHSSRHRR | |
Greater than 80% as determined by SDS-PAGE. |
59086 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |