missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat S100A8 / CAGA Protein

A DNA sequence encoding the Rattus norvegicus (Rat) S100A8 / CAGA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6642-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044750

  • 675.00 EUR / 100µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_446274.2
S100A8 / CAGA Protein
Research Use Only
S100A8
Recombinant Protein
E. coli
ATELEKALSN VIEVYHNYSG IKGNHHALYR DDFRKMVTTE CPQFVQNKNT ESLFKELDVN SDNAINFEEF LVLVIRVGVA AHKDSHKE
Greater than 90% as determined by SDS-PAGE.
116547
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers