missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat S100A8 / CAGA Protein
A DNA sequence encoding the Rattus norvegicus (Rat) S100A8 / CAGA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6642-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_446274.2 | |
S100A8 / CAGA Protein | |
Research Use Only | |
S100A8 | |
Recombinant Protein | |
E. coli | |
ATELEKALSN VIEVYHNYSG IKGNHHALYR DDFRKMVTTE CPQFVQNKNT ESLFKELDVN SDNAINFEEF LVLVIRVGVA AHKDSHKE | |
Greater than 90% as determined by SDS-PAGE. |
116547 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |